Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145387.4 | NP_663362.2 | 10 | 1649 | ||
GenBank mRNA | AK077662.1 | - | 10 | 1650 | ||
AK085600.1 | - | 9 | 1820 | |||
AK220240.1 | - | 6 | 2881 | |||
BC023151.1 | - | 9 | 1335 | |||
BC056953.1 | - | 10 | 1611 | 5717810 |