Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012474.4 | NP_036606.2 | 5 | 874 | ||
XM_011509970.2 | XP_011508272.2 | 3 | 409 | |||
XM_017002236.1 | XP_016857725.1 | 6 | 1079 | |||
GenBank mRNA | AB062451.1 | - | 5 | 493 | ||
AF236637.1 | - | 5 | 553 | |||
AK294537.1 | - | 4 | 404 | |||
BC002906.2 | - | 5 | 714 | 3940564 | ||
BT006860.1 | - | 2 | 85 | |||
CR456857.1 | - | 2 | 85 | |||
D78335.1 | - | 3 | 303 |