Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cplx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145493.1 | NP_663468.1 | 1 | 254 | ||
GenBank mRNA | AK044262.1 | - | 1 | 262 | ||
AY264291.1 | - | 1 | 143 | |||
BC016082.1 | - | 1 | 254 | 4501240 | ||
BC029011.1 | - | 1 | 202 | 5357047 |