Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145494.2 | NP_663469.1 | 2 | 179 | ||
GenBank mRNA | AK039906.1 | - | 2 | 226 | ||
AK042289.1 | - | 2 | 226 | |||
AK043103.1 | - | 2 | 217 | |||
AK050933.1 | - | 2 | 226 | |||
AK050980.1 | - | 2 | 226 | |||
AK077746.1 | - | 2 | 226 | |||
AK080403.1 | - | 2 | 237 | |||
AK156403.1 | - | 2 | 226 | |||
AK162126.1 | - | 2 | 238 | |||
AK171157.1 | - | 2 | 226 | |||
BC004709.1 | - | 2 | 179 | 3499365 |