Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5430435G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145509.2 | NP_663484.1 | 3 | 269 | ||
GenBank mRNA | AB232641.1 | - | 2 | 113 | ||
AK030688.1 | - | 3 | 269 | |||
AK036056.1 | - | 3 | 258 | |||
AK079999.1 | - | 3 | 237 | |||
CJ137398.1 | - | 3 | 188 | |||
JX431303.1 | - | 2 | 113 |