Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145510.1 | NP_663485.1 | 1 | 132 | ||
GenBank mRNA | AK087547.1 | - | 1 | 127 | ||
AK133528.1 | - | 1 | 153 | |||
AK155054.1 | - | 1 | 132 | |||
AK156126.1 | - | 44 | ||||
AK158129.1 | - | 145 | ||||
AK165748.1 | - | 1 | 150 | |||
AK165795.1 | - | 145 | ||||
AK207658.1 | - | 1 | 50 | |||
BC011166.1 | - | 1 | 132 | 4206804 | ||
BC024808.1 | - | 1 | 122 | 5356768 |