Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145517.4 | NP_663492.3 | 3 | 351 | ||
GenBank mRNA | AK050385.1 | - | 3 | 336 | ||
AK077562.1 | - | 3 | 334 | |||
BC012315.1 | - | 3 | 306 | 3981845 | ||
BC023695.1 | - | 3 | 331 | 5344622 | ||
BC023903.1 | - | 3 | 294 | |||
BC025572.1 | - | 3 | 327 | 5323819 | ||
CN705165.1 | - | 3 | 351 |