Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 2 | 230 | ||
GenBank mRNA | AK050860.1 | - | 2 | 190 | ||
AK086699.1 | - | 2 | 223 | |||
AK171713.1 | - | 2 | 256 | |||
BB663505.1 | - | 2 | 223 | |||
BC009153.1 | - | 2 | 219 | 2655209 | ||
BY130680.1 | - | 2 | 231 |