Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145524.3 | NP_663499.2 | 4 | 582 | ||
GenBank mRNA | AK042669.1 | - | 4 | 560 | ||
AK087927.1 | - | 4 | 512 | |||
BC004636.1 | - | 4 | 503 | 3585719 | ||
BC057960.1 | - | 4 | 449 | 5354784 | ||
DQ093357.1 | - | 4 | 559 | |||
DQ093358.1 | - | 4 | 559 | |||
DQ230973.1 | - | 3 | 350 |