Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145546.1 | NP_663521.1 | 4 | 390 | ||
GenBank mRNA | AK131856.1 | - | 4 | 412 | ||
AK133097.1 | - | 4 | 412 | |||
AK133825.1 | - | 4 | 412 | |||
AK143935.1 | - | 4 | 412 | |||
AK145403.1 | - | 4 | 412 | |||
AK146223.1 | - | 4 | 413 | |||
AK146666.1 | - | 4 | 412 | |||
AK146910.1 | - | 4 | 412 | |||
AK151577.1 | - | 4 | 412 | |||
BC016637.1 | - | 4 | 390 | 2650779 |