Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170540.1 | NP_001164011.1 | 3, 4 | 332 | ||
NM_145455.3 | NP_663430.2 | 3, 4 | 534 | |||
GenBank mRNA | AK139664.1 | - | 3, 4 | 307 | ||
AK146332.1 | - | 3, 4 | 320 | |||
AK147171.1 | - | 3, 4 | 307 | |||
AK160227.1 | - | 3, 4 | 306 | |||
BC008233.1 | - | 3, 4 | 293 | 2648048 | ||
BC064010.1 | - | 3, 4 | 534 | 30244324 | ||
BC080837.1 | - | 3, 4 | 288 | 5371708 | ||
CJ073405.1 | - | 3, 4 | 333 |