Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145476.2 | NP_663451.2 | 7, 8 | 1012 | ||
GenBank mRNA | AK039627.1 | - | 7, 8 | 1025 | ||
AK146872.1 | - | 7, 8 | 1013 | |||
AK154226.1 | - | 7, 8 | 1034 | |||
AK154602.1 | - | 7, 8 | 1055 | |||
AK155459.1 | - | 7, 8 | 1026 | |||
AK155770.1 | - | 7, 8 | 1055 | |||
AK216948.1 | - | 3, 4 | 237 | |||
BC023106.1 | - | 4, 5 | 407 | 4016550 | ||
BC066009.1 | - | 7, 8 | 1012 | 5708486 |