Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000667.3 | NP_000658.1 | 7, 8 | 1070 | ||
GenBank mRNA | AK290558.1 | - | 7, 8 | 1027 | ||
BC074738.2 | - | 7, 8 | 984 | 30915195 | ||
BC117442.1 | - | 7, 8 | 1026 | 40125993 | ||
BC126306.1 | - | 7, 8 | 1026 | 8992022 | ||
BG199182.1 | - | 1, 2 | 133 | |||
BT019812.1 | - | 7, 8 | 956 | |||
M12271.1 | - | 7, 8 | 971 | |||
M12963.1 | - | 7, 8 | 1028 |