Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164359.1 | NP_001157831.1 | 7, 8 | 710 | ||
NM_001164360.1 | NP_001157832.1 | 7, 8 | 618 | |||
NM_145502.3 | NP_663477.3 | 6, 7 | 814 | |||
GenBank mRNA | AK133557.1 | - | 6, 7 | 557 | ||
AK145990.1 | - | 7, 8 | 591 | |||
BC011220.1 | - | 7, 8 | 582 | 4208238 | ||
BC023849.1 | - | 6, 7 | 677 | 5345801 |