Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 9 | 1248 | ||
NM_001286650.1 | NP_001273579.1 | 10 | 1370 | |||
GenBank mRNA | AF153821.1 | - | 9 | 1205 | ||
AK299409.1 | - | 10 | 1166 | |||
AK301018.1 | - | 11 | 1308 | |||
AK303945.1 | - | 8 | 1038 | |||
AK304396.1 | - | 9 | 972 | |||
AK316501.1 | - | 9 | 1038 | |||
BC033009.2 | - | 9 | 1207 | 4828235 | ||
BX647469.1 | - | 9 | 3232 | |||
D00137.1 | - | 9 | 1157 | |||
FJ224285.1 | - | 9 | 1207 | |||
M21692.1 | - | 9 | 1169 | |||
M24317.1 | - | 9 | 1167 | |||
X03350.1 | - | 9 | 1209 |