Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145516.2 | NP_663491.1 | 7 | 584 | ||
NM_175421.1 | NP_780630.1 | 7 | 584 | |||
GenBank mRNA | AF189817.1 | - | 7 | 544 | ||
AK039771.1 | - | 7 | 552 | |||
AK082118.1 | - | 7 | 537 | |||
AK148653.1 | - | 7 | 567 | |||
AK149868.1 | - | 7 | 544 | |||
AK150074.1 | - | 7 | 544 | |||
AK166091.1 | - | 7 | 552 | |||
BC057994.1 | - | 7 | 552 | 5344583 | ||
CK625615.1 | - | 7 | 584 |