Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 3 | 318 | ||
GenBank mRNA | AK050860.1 | - | 3 | 278 | ||
AK086699.1 | - | 3 | 311 | |||
AK171713.1 | - | 3 | 344 | |||
BB663505.1 | - | 3 | 311 | |||
BC009153.1 | - | 3 | 307 | 2655209 |