Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290733.1 | NP_001277662.1 | 10, 11 | 1529 | ||
NM_001290734.1 | NP_001277663.1 | 9, 10 | 1289 | |||
NM_145525.3 | NP_663500.2 | 9, 10 | 1289 | |||
GenBank mRNA | AK037703.1 | - | 9, 10 | 1240 | ||
AK044411.1 | - | 10, 11 | 1500 | |||
AK220498.1 | - | 9, 10 | 1236 | |||
BC022908.1 | - | 4, 5 | 448 | 5043261 |