Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Madd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177719.1 | NP_001171190.1 | 3 | 662 | ||
NM_001177720.1 | NP_001171191.1 | 2 | 410 | |||
NM_145527.4 | NP_663502.3 | 3 | 662 | |||
GenBank mRNA | AK035777.1 | - | 3 | 641 | ||
AK036347.1 | - | 3 | 663 | |||
AK122257.1 | - | 3 | 504 | |||
AY263980.1 | - | 3 | 480 | |||
AY263981.1 | - | 3 | 480 | |||
AY263982.1 | - | 3 | 480 | |||
AY263983.1 | - | 3 | 480 | |||
AY263984.1 | - | 3 | 480 | |||
AY263985.1 | - | 3 | 480 | |||
AY263986.1 | - | 3 | 480 | |||
AY263987.1 | - | 3 | 480 | |||
AY263988.1 | - | 3 | 480 | |||
AY263989.1 | - | 3 | 480 | |||
BC003255.1 | - | 3 | 499 | |||
BC063386.1 | - | 3 | 463 | 5401819 |