Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 6 | 1512 | ||
GenBank mRNA | AK080983.1 | - | 1653 | |||
AK088196.1 | - | 6 | 1524 | |||
AK151326.1 | - | 1498 | ||||
AK151705.1 | - | 6 | 1522 | |||
AK180745.1 | - | 1 | 217 | |||
AK187561.1 | - | 1 | 135 | |||
AK197493.1 | - | 1 | 182 | |||
AK213035.1 | - | 1 | 150 | |||
BC002270.1 | - | 6 | 1512 | 3496088 |