Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9030617O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145448.4 | NP_663423.2 | 10 | 1575 | ||
GenBank mRNA | AK033551.1 | - | 10 | 1604 | ||
AK038436.1 | - | 10 | 1577 | |||
AK081256.1 | - | 1612 | ||||
AK152622.1 | - | 10 | 1604 | |||
BC021385.1 | - | 11 | 1560 | 5064508 | ||
BC066161.1 | - | 11 | 1649 | 30093441 |