Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000673.4 | NP_000664.2 | 3 | 492 | ||
NM_001166504.1 | NP_001159976.1 | 3 | 305 | |||
GenBank mRNA | AK223472.1 | - | 3 | 286 | ||
AK301696.1 | - | 3 | 305 | |||
AK312854.1 | - | 3 | 415 | |||
BC131512.1 | - | 3 | 287 | 40083994 | ||
BX476080.1 | - | 3 | 349 | |||
CD014081.1 | - | 4 | 368 | |||
L33179.1 | - | 3 | 215 | |||
U07821.1 | - | 3 | 299 | |||
U09623.1 | - | 3 | 233 | |||
X76342.1 | - | 3 | 275 |