Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145452.3 | NP_663427.2 | 9 | 1860 | ||
GenBank mRNA | AK133354.1 | - | 5 | 420 | ||
BC013637.1 | - | 7 | 606 | 3498774 | ||
BC065082.1 | - | 9 | 1403 |