Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145479.4 | NP_663454.3 | 7 | 2497 | ||
GenBank mRNA | AK028197.1 | - | 8 | 2642 | ||
AK143514.1 | - | 7 | 2442 | |||
AK190702.1 | - | 1 | 190 | |||
AK191360.1 | - | 1 | 190 | |||
AK204888.1 | - | 1 | 191 | |||
AK211998.1 | - | 1 | 129 | |||
BC005800.1 | - | 7 | 2475 | 2812589 |