Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145492.4 | NP_663467.1 | 8 | 4228 | ||
GenBank mRNA | AK052655.1 | - | 8 | 4132 | ||
AK077697.1 | - | 5 | 1050 | |||
AK081096.1 | - | 6 | 1682 | |||
AK131130.1 | - | 7 | 4313 | |||
AY147406.1 | - | 8 | 4131 | |||
BC021376.1 | - | 8 | 4044 | 5038671 | ||
BC023458.1 | - | 5 | 764 |