Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145495.2 | NP_663470.1 | 10 | 3967 | ||
GenBank mRNA | BC011277.1 | - | 10 | 3967 | 3711198 |