Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000690.3 | NP_000681.2 | 2, 3 | 323 | ||
GenBank mRNA | AK223373.1 | - | 2, 3 | 265 | ||
AK314856.1 | - | 2, 3 | 245 | |||
AY621070.1 | - | 2, 3 | 288 | |||
BC002967.1 | - | 2, 3 | 273 | 3543343 | ||
BC071839.1 | - | 2, 3 | 232 | 5477768 | ||
CR456991.1 | - | 2, 3 | 227 | |||
X05409.1 | - | 2, 3 | 260 | |||
Y00109.1 | - | 2, 3 | 251 |