Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC003331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077237.1 | NP_001070705.1 | 13 | 1821 | ||
NM_145511.2 | NP_663486.2 | 14 | 1917 | |||
GenBank mRNA | AK152022.1 | - | 11 | 1337 | ||
AK152955.1 | - | 11 | 1338 | |||
BC003331.1 | - | 12 | 1506 | 3256634 |