Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppapdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145521.3 | NP_663496.1 | 2 | 1210 | ||
GenBank mRNA | AK048016.1 | - | 3 | 838 | ||
AK052883.1 | - | 2 | 1211 | |||
AK134638.1 | - | 2 | 1334 | |||
BC016136.1 | - | 2 | 1141 | 4910887 |