Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145530.2 | NP_663505.1 | 3 | 1609 | ||
GenBank mRNA | AK036775.1 | - | 3 | 1610 | ||
AK037170.1 | - | 3 | 1611 | |||
BC021307.1 | - | 3 | 1614 | 5003249 |