Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 6 | 1546 | ||
GenBank mRNA | AK080983.1 | - | 1687 | |||
AK088196.1 | - | 6 | 1558 | |||
AK151326.1 | - | 1532 | ||||
AK151705.1 | - | 6 | 1556 | |||
AK170042.1 | - | 1559 | ||||
AK180745.1 | - | 1 | 251 | |||
AK187561.1 | - | 1 | 169 | |||
AK197493.1 | - | 1 | 216 | |||
BC002270.1 | - | 6 | 1546 | 3496088 |