Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145539.2 | NP_663514.2 | 5 | 1357 | ||
GenBank mRNA | AK148484.1 | - | 5 | 1326 | ||
BC010814.1 | - | 5 | 1308 | 4211816 |