Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145541.5 | NP_663516.1 | 5 | 381 | ||
GenBank mRNA | AK030897.1 | - | 5 | 331 | ||
AK152504.1 | - | 5 | 301 | |||
BC051419.1 | - | 5 | 337 | 3494423 | ||
BC083128.1 | - | 4 | 233 | 6438722 | ||
CX206795.1 | - | 5 | 294 | |||
EU359271.1 | - | 4 | 243 |