Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000695.3 | NP_000686.2 | 9 | 1374 | ||
NM_001031615.1 | NP_001026786.1 | 9 | 1218 | |||
GenBank mRNA | AK092464.1 | - | 9 | 1431 | ||
AK299956.1 | - | 4 | 699 | |||
BC007685.2 | - | 9 | 1218 | 3636326 | ||
BT006810.1 | - | 7 | 935 |