Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000701.7 | NP_000692.2 | 2 | 422 | ||
NM_001160233.1 | NP_001153705.1 | 2 | 446 | |||
NM_001160234.1 | NP_001153706.1 | 2 | 255 | |||
XM_017001360.1 | XP_016856849.1 | 3 | 3641 | |||
XM_017001361.1 | XP_016856850.1 | 3 | 2983 | |||
GenBank mRNA | AK295095.1 | - | 2 | 445 | ||
AK296362.1 | - | 2 | 255 | |||
AK296384.1 | - | 2 | 381 | |||
AK308418.1 | - | 2 | 187 | |||
AK314777.1 | - | 2 | 267 | |||
BC003077.2 | - | 2 | 335 | 3507685 | ||
BC050359.1 | - | 2 | 333 | 6065209 | ||
BF795089.1 | - | 382 | ||||
DC335233.1 | - | 2 | 422 | |||
U16798.1 | - | 1 | 83 |