Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145576.2 | NP_663551.2 | 4, 5 | 776 | ||
GenBank mRNA | BC006693.1 | - | 4, 5 | 661 | 3582679 | |
BC088737.1 | - | 4, 5 | 658 | 5372342 | ||
CF915685.1 | - | 604 |