Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 2, 3 | 245 | ||
GenBank mRNA | AB018321.2 | - | 2, 3 | 202 | ||
AK091617.1 | - | 2, 3 | 216 | |||
AK314296.1 | - | 2, 3 | 216 | |||
AL831991.1 | - | 1, 2 | 34 | |||
AL831997.1 | - | 1, 2 | 60 | |||
BC052271.1 | - | 2, 3 | 196 | 6672133 | ||
DA221602.1 | - | 2, 3 | 245 | |||
KU177927.1 | - | 1, 2 | 113 | |||
KU177928.1 | - | 1, 2 | 113 |