Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145584.2 | NP_663559.1 | 1 | 538 | ||
GenBank mRNA | AK134774.1 | - | 1 | 363 | ||
AK146506.1 | - | 1 | 363 | |||
AK173021.1 | - | 1 | 205 | |||
BC020531.1 | - | 1 | 408 | 4221758 |