Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195413.1 | NP_001182342.1 | 8 | 669 | ||
NM_145601.1 | NP_663576.1 | 8 | 669 | |||
GenBank mRNA | AK080705.1 | - | 8 | 656 | ||
BC016201.1 | - | 8 | 669 | 4527347 | ||
BC051415.1 | - | 6 | 1578 | |||
BC058272.1 | - | 7 | 621 | |||
HQ116386.1 | - | 8 | 669 |