Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195006.1 | NP_001181935.1 | 3 | 299 | ||
NM_145602.3 | NP_663577.1 | 3 | 299 | |||
GenBank mRNA | AK090374.1 | - | 3 | 281 | ||
AK144294.1 | - | 3 | 281 | |||
AK168873.1 | - | 3 | 283 | |||
BC006595.1 | - | 3 | 273 | 3156802 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129305.1 | - | 594 | Chr. - - [-] on Build |