Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D230025D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145604.2 | NP_663579.2 | 2 | 372 | ||
GenBank mRNA | AK051965.1 | - | 2 | 358 | ||
AK156120.1 | - | 2 | 374 | |||
AK166937.1 | - | 2 | 355 | |||
AK218692.1 | - | 1 | 15 | |||
BC006874.1 | - | 2 | 255 | 2646515 | ||
BC057919.1 | - | 2 | 361 | 5134302 | ||
BC066141.1 | - | 2 | 293 | |||
BC066826.1 | - | 2 | 293 | |||
CN703950.1 | - | 2 | 372 |