Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145615.4 | NP_663590.3 | 2 | 327 | ||
GenBank mRNA | AK028118.1 | - | 2 | 328 | ||
AK032830.1 | - | 2 | 280 | |||
AK133525.1 | - | 2 | 273 | |||
AK167572.1 | - | 2 | 300 | |||
AK168321.1 | - | 2 | 288 | |||
AK218235.1 | - | 1 | 18 | |||
BC003432.1 | - | 2 | 218 | 2646522 | ||
BC096645.1 | - | 2 | 186 | 1245213 |