Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ice2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145618.3 | NP_663593.2 | 10, 11 | 2299 | ||
GenBank mRNA | AK134186.1 | - | 10, 11 | 2344 | ||
AK163917.1 | - | 10, 11 | 2227 | |||
AY244558.1 | - | 10, 11 | 2328 | |||
BC018458.1 | - | 1, 2 | 650 | |||
BC068183.1 | - | 10, 11 | 2299 | 30639735 | ||
BC080786.1 | - | 1, 2 | 940 |