Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145619.2 | NP_663594.2 | 3 | 360 | ||
GenBank mRNA | AF368233.1 | - | 3 | 360 | ||
AK044223.1 | - | 3 | 338 | |||
AK088125.1 | - | 3 | 362 | |||
AK145259.1 | - | 3 | 379 | |||
AK147868.1 | - | 3 | 659 | |||
AK165070.1 | - | 3 | 290 | |||
AK170541.1 | - | 3 | 304 | |||
AK177929.1 | - | 1 | 27 | |||
BC014870.1 | - | 3 | 300 | 3602116 | ||
BC058754.1 | - | 3 | 280 | 6476689 | ||
BC062204.1 | - | 3 | 280 |