Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145625.3 | NP_663600.2 | 10 | 1425 | ||
GenBank mRNA | AK045250.1 | - | 10 | 1269 | ||
AK077423.1 | - | 10 | 1276 | |||
AK148012.1 | - | 10 | 1275 | |||
AK161693.1 | - | 10 | 1258 | |||
AK163266.1 | - | 10 | 1276 | |||
AK166363.1 | - | 10 | 1277 | |||
AK168350.1 | - | 10 | 1271 | |||
AK168425.1 | - | 10 | 1271 | |||
AK170256.1 | - | 10 | 1276 | |||
AK170354.1 | - | 10 | 1258 | |||
AK178737.1 | - | 1 | 47 | |||
AK186828.1 | - | 1 | 38 | |||
AK206900.1 | - | 1 | 47 | |||
AK214347.1 | - | 1 | 47 | |||
BC007171.1 | - | 9 | 1224 | 3492114 | ||
BC036293.1 | - | 10 | 1261 |