Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMF1-BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199661.1 | NP_001186590.1 | 7 | 828 | ||
NM_001199662.1 | NP_001186591.1 | 7 | 889 | |||
NM_001199663.1 | NP_001186592.1 | 6 | 693 | |||
NM_001199664.1 | NP_001186593.1 | 5 | 682 | |||
GenBank mRNA | BC033656.2 | - | 6 | 683 |
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199173.5 | NP_954642.1 | 4 | 631 | ||
GenBank mRNA | BC113432.1 | - | 4 | 433 | 8322484 | |
BC113434.1 | - | 4 | 433 | 8322486 | ||
X51699.1 | - | 4 | 379 | |||
X53698.1 | - | 4 | 379 |