Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145709.2 | NP_663755.2 | 2 | 303 | ||
GenBank mRNA | BC052664.1 | - | 2 | 307 | 30053785 | |
CJ052752.1 | - | 2 | 305 |
Gene Symbol: Obox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145707.3 | NP_663753.3 | 3 | 310 | ||
GenBank mRNA | AK139750.1 | - | 3 | 311 | ||
AY094981.1 | - | 700 | ||||
BC119564.1 | - | 1 | 572 |
Gene Symbol: Obox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038676.1 | NP_001033765.1 | 2 | 262 | ||
GenBank mRNA | BC099488.1 | - | 2 | 262 | 30939375 |