Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291144.1 | NP_001278073.1 | 5 | 462 | ||
NM_145742.2 | NP_665685.1 | 5 | 442 | |||
GenBank mRNA | AK004665.1 | - | 5 | 388 | ||
AK155482.1 | - | 5 | 381 | |||
AK156955.1 | - | 5 | 408 | |||
AK170952.1 | - | 5 | 381 | |||
BC029709.1 | - | 5 | 429 | 5359253 |