Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145588.1 | NP_663563.1 | 3 | 364 | ||
GenBank mRNA | AK044826.1 | - | 2 | 1823 | ||
AK075922.1 | - | 3 | 355 | |||
AK084310.1 | - | 3 | 352 | |||
BC003427.1 | - | 3 | 364 | 2615715 |