Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145595.2 | NP_663570.2 | 3 | 498 | ||
GenBank mRNA | AK031756.1 | - | 2 | 1940 | ||
AK043313.1 | - | 3 | 323 | |||
AK150763.1 | - | 3 | 499 | |||
BC009118.1 | - | 3 | 448 | 3154595 |